ENA258389386 Enamine

2-(1H-1,3-benzodiazol-1-yl)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD14705796
  • Cas Number : 1221725-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59614
Availability details for
Quantity
  • Availability
  • Add to Cart