ENA258389388 Enamine

methyl[3-(3-phenyl-1,2-oxazol-5-yl)propyl]amine

  • MDL number MFCD14705799
  • Cas Number : 1221722-20-8
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59626
Availability details for
Quantity
  • Availability
  • Add to Cart