ENA258389390 Enamine

4-{[3-(aminomethyl)benzene]sulfonyl}piperazin-2-one hydrochloride

  • MDL number MFCD14705802
  • Cas Number : 1235440-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59659
Availability details for
Quantity
  • Availability
  • Add to Cart