ENA258389391 Enamine

7-amino-2,4-dimethylpyrimido[2,1-a]phthalazin-5-ium chloride

  • MDL number MFCD14705803
  • Cas Number : 54752-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 332...334
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59660

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59660
Availability details for
Quantity
  • Availability
  • Add to Cart