ENA258389405 Enamine

3-[(2,4-dioxo-1,3-thiazolidin-3-yl)methyl]benzonitrile

  • MDL number MFCD14705819
  • Cas Number : 1221723-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59777
Availability details for
Quantity
  • Availability
  • Add to Cart