ENA258389407 Enamine

N-[1-(1-methyl-1H-pyrazol-4-yl)ethylidene]hydroxylamine

  • Cas Number : 1221728-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59788
Availability details for
Quantity
  • Availability
  • Add to Cart