ENA258389408 Enamine

3-[4-(2-hydroxyethyl)-3-(methoxycarbonyl)-5-oxo-4,5-dihydro-1H-pyrazol-1-yl]benzoic acid

  • MDL number MFCD14705822
  • Cas Number : 1221723-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59790
Availability details for
Quantity
  • Availability
  • Add to Cart