ENA258389410 Enamine

2-[({3-bromoimidazo[1,2-a]pyridin-2-yl}methyl)sulfanyl]benzoic acid hydrobromide

  • MDL number MFCD14705825
  • Cas Number : 1221723-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59800
Availability details for
Quantity
  • Availability
  • Add to Cart