ENA258389413 Enamine

1-(3,4-dichlorophenyl)butan-1-amine hydrochloride

  • MDL number MFCD14705828
  • Cas Number : 90944-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 316...318
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59839

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59839
Availability details for
Quantity
  • Availability
  • Add to Cart