ENA258389414 Enamine

2-(pyridin-3-yloxy)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD14705829
  • Cas Number : 1221722-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59854
Availability details for
Quantity
  • Availability
  • Add to Cart