ENA258389421 Enamine

3-(pyridin-3-yl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD14705837
  • Cas Number : 1071623-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59901

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59901
Availability details for
Quantity
  • Availability
  • Add to Cart