ENA258389424 Enamine

2-(2,3-dihydro-1H-inden-5-yloxy)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD14705840
  • Cas Number : 1223627-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59920
Availability details for
Quantity
  • Availability
  • Add to Cart