ENA258389425 Enamine

N-(5-fluoro-2-methylphenyl)-2-(N-hydroxyimino)acetamide

  • MDL number MFCD14705841
  • Cas Number : 1846567-77-8
  • Molecular Weight : 196.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59922
Availability details for
Quantity
  • Availability
  • Add to Cart