ENA258389427 Enamine

{[3-methoxy-4-(methylsulfanyl)phenyl]methyl}(methyl)amine

  • MDL number MFCD14705843
  • Cas Number : 1221723-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59938

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59938
Availability details for
Quantity
  • Availability
  • Add to Cart