ENA258389430 Enamine

2-(4-bromophenyl)-1,3-thiazole-4-sulfonyl chloride

  • MDL number MFCD14705846
  • Cas Number : 1221723-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59959

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59959
Availability details for
Quantity
  • Availability
  • Add to Cart