ENA258389431 Enamine

2,2,2-trifluoroethyl N-[3-fluoro-4-(4-methyl-1,4-diazepan-1-yl)phenyl]carbamate hydrochloride

  • MDL number MFCD14705847
  • Cas Number : 1221724-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59961
Availability details for
Quantity
  • Availability
  • Add to Cart