ENA258389439 Enamine

1-[4-(difluoromethane)sulfonyl-3-methoxyphenyl]ethan-1-one

  • MDL number MFCD14705857
  • Cas Number : 1235441-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60013
Availability details for
Quantity
  • Availability
  • Add to Cart