ENA258389440 Enamine

(2E)-3-(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)prop-2-enoic acid

  • MDL number MFCD14705858
  • Cas Number : 68032-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60017

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60017
Availability details for
Quantity
  • Availability
  • Add to Cart