ENA258389446 Enamine

4-({[(1-ethylpyrrolidin-2-yl)methyl]amino}methyl)phenol dihydrochloride

  • MDL number MFCD14705864
  • Cas Number : 1235439-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60075
Availability details for
Quantity
  • Availability
  • Add to Cart