ENA258389452 Enamine

(5-chloro-1-methyl-1H-imidazol-2-yl)methanol hydrochloride

  • MDL number MFCD14705872
  • Cas Number : 1235441-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60137
Availability details for
Quantity
  • Availability
  • Add to Cart