ENA258389457 Enamine

1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD14705878
  • Cas Number : 1235439-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60230

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60230
Availability details for
Quantity
  • Availability
  • Add to Cart