ENA258389525 Enamine

2-(2-methyl-1H-imidazol-1-yl)quinoline-4-carboxylic acid

  • MDL number MFCD12652350
  • Cas Number : 1097017-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59441
Availability details for
Quantity
  • Availability
  • Add to Cart