ENA258389528 Enamine

4-[(1H-1,3-benzodiazol-1-yl)methyl]aniline

  • MDL number MFCD09742949
  • Cas Number : 142335-67-9
  • Molecular Weight : 223.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59458
Availability details for
Quantity
  • Availability
  • Add to Cart