ENA258389550 Enamine

2-(2,4-dimethylphenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07375285
  • Cas Number : 886368-73-6
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59541
Availability details for
Quantity
  • Availability
  • Add to Cart