ENA258389556 Enamine

3-ethylmorpholine

  • MDL number MFCD09054774
  • Cas Number : 55265-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43359
Availability details for
Quantity
  • Availability
  • Add to Cart