ENA258389561 Enamine

5-(3-chlorophenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD05182336
  • Cas Number : 168893-31-0
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-48626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48626
Availability details for
Quantity
  • Availability
  • Add to Cart