ENA258389564 Enamine

{4-[(thiophen-2-yl)methoxy]phenyl}methanol

  • MDL number MFCD07359796
  • Cas Number : 881447-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59576
Availability details for
Quantity
  • Availability
  • Add to Cart