ENA258389573 Enamine

(methylsulfamoyl)amine

  • MDL number MFCD02691960
  • Cas Number : 72179-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50354
Availability details for
Quantity
  • Availability
  • Add to Cart