ENA258389575 Enamine

2-{[1-(3-ethyl-1-benzofuran-2-yl)ethyl]amino}acetamide

  • MDL number MFCD09050891
  • Cas Number : 926231-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59590

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59590
Availability details for
Quantity
  • Availability
  • Add to Cart