ENA258389576 Enamine

1-(propane-1-sulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD03830447
  • Cas Number : 854137-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59592
Availability details for
Quantity
  • Availability
  • Add to Cart