ENA258389578 Enamine

4-(methanesulfonylmethyl)aniline

  • MDL number MFCD11846977
  • Cas Number : 24176-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59599
Availability details for
Quantity
  • Availability
  • Add to Cart