ENA258389583 Enamine

1,1-dioxo-1lambda6-thiolan-3-yl 4-methylbenzene-1-sulfonate

  • MDL number MFCD00434940
  • Cas Number : 36715-85-2
  • Molecular Weight : 290.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59604
Availability details for
Quantity
  • Availability
  • Add to Cart