ENA258389586 Enamine

2-(2-methylphenyl)-1H-1,3-benzodiazole

  • MDL number MFCD00453934
  • Cas Number : 2963-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16232
Availability details for
Quantity
  • Availability
  • Add to Cart