ENA258389590 Enamine

2-(ethylamino)-2-methylpropan-1-ol

  • MDL number MFCD11053962
  • Cas Number : 82922-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59613

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59613
Availability details for
Quantity
  • Availability
  • Add to Cart