ENA258389592 Enamine

3-[(6-aminopyridin-3-yl)oxy]benzamide

  • MDL number MFCD11135854
  • Cas Number : 1019634-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59615
Availability details for
Quantity
  • Availability
  • Add to Cart