ENA258389620 Enamine

N-benzyl-N-methyl-5-nitropyridin-2-amine

  • MDL number MFCD05693829
  • Cas Number : 181114-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59662
Availability details for
Quantity
  • Availability
  • Add to Cart