ENA258389623 Enamine

1-benzyl-4-(2,5-dimethyl-1H-pyrrol-1-yl)piperidine

  • MDL number MFCD13680187
  • Cas Number : 1108147-59-6
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59665
Availability details for
Quantity
  • Availability
  • Add to Cart