ENA258389625 Enamine

methyl 2-amino-2-(4-bromophenyl)acetate hydrochloride

  • MDL number MFCD05663802
  • Cas Number : 42718-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59667
Availability details for
Quantity
  • Availability
  • Add to Cart