ENA258389627 Enamine

5-(2,5-dimethylphenyl)-1,2,4-triazin-3-amine

  • MDL number MFCD11134076
  • Cas Number : 1094385-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59670
Availability details for
Quantity
  • Availability
  • Add to Cart