ENA258389628 Enamine

methyl 3-(3-oxobutanamido)benzoate

  • MDL number MFCD09804436
  • Cas Number : 1016715-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59671

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59671
Availability details for
Quantity
  • Availability
  • Add to Cart