ENA258389629 Enamine

2-[(3-ethynylphenyl)amino]-N-methylacetamide

  • MDL number MFCD11149845
  • Cas Number : 1020974-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59672
Availability details for
Quantity
  • Availability
  • Add to Cart