ENA258389633 Enamine

1-(4-iodophenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD05174727
  • Cas Number : 773873-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59676

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59676
Availability details for
Quantity
  • Availability
  • Add to Cart