ENA258389646 Enamine

heptane-4-thiol

  • MDL number MFCD11193830
  • Cas Number : 1639-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59690
Availability details for
Quantity
  • Availability
  • Add to Cart