ENA258389652 Enamine

(5-methyl-1H-1,3-benzodiazol-2-yl)(phenyl)methanol

  • MDL number MFCD11106900
  • Cas Number : 18046-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59708
Availability details for
Quantity
  • Availability
  • Add to Cart