ENA258389657 Enamine

1-benzyl-2-ethyl-4,5,6,7-tetrahydro-1H-indol-4-one

  • MDL number MFCD14705810
  • Cas Number : 218934-50-0
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59715
Availability details for
Quantity
  • Availability
  • Add to Cart