ENA258389659 Enamine

4-(2-oxopyrrolidin-1-yl)benzene-1-sulfonamide

  • MDL number MFCD00453080
  • Cas Number : 36090-27-4
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59717
Availability details for
Quantity
  • Availability
  • Add to Cart