ENA258389665 Enamine

3-[(tert-butylsulfamoyl)methyl]benzoic acid

  • MDL number MFCD11550779
  • Cas Number : 1098373-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59723
Availability details for
Quantity
  • Availability
  • Add to Cart