ENA258389667 Enamine

1,4-oxazepane

  • MDL number MFCD09971804
  • Cas Number : 5638-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59725

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-59725
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart