ENA258389668 Enamine

2,3-dihydro-1H-indene-1-thiol

  • MDL number MFCD10689662
  • Cas Number : 121607-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59730
Availability details for
Quantity
  • Availability
  • Add to Cart