ENA258389669 Enamine

1-(4-methoxyphenyl)ethane-1-thiol

  • MDL number MFCD10694699
  • Cas Number : 113682-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59732

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59732
Availability details for
Quantity
  • Availability
  • Add to Cart